5-Phenylthieno[2,3-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

A109803-1G A109803-250MG A109803-25G A109803-5G
76869-664EA 28.65 USD
76869-664 76869-666 76869-668 76869-670
5-Phenylthieno[2,3-d]pyrimidin-4(3H)-one 98%
5-Phenylthieno[2,3-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₂H₈N₂OS
MW: 228.27 g/mol
MDL Number: MFCD00126441
CAS Number: 35978-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR