L-Ascorbic acid 2-glucoside 97%

Supplier: Ambeed
Warning

Synonyms: 2-O-α-D-Glucopyranosyl-L-ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-4-hydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)furan-2(5H)-one

A109609-10G A109609-1G A109609-25G A109609-5G A109609-100G
76869-656EA 16.98 USD
76869-656 76869-658 76869-660 76869-662 77670-550
L-Ascorbic acid 2-glucoside 97%
L-Ascorbic acid 2-glucoside

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₁₁
MW: 338.26 g/mol
MDL Number: MFCD00925623
CAS Number: 129499-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR