6-Phenylbenzo[d]isoxazol-3-amine 98%

Supplier: Ambeed
Warning

A109523-1G A109523-250MG
76869-652EA 341.61 USD
76869-652 76869-654
6-Phenylbenzo[d]isoxazol-3-amine 98%
6-Phenylbenzo[d]isoxazol-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O
MW: 210.23 g/mol
MDL Number: MFCD13193391
CAS Number: 268734-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR