4,5-Difluorobenzene-1,2-diol 97%

Supplier: Ambeed
Warning

A109497-100MG A109497-1G A109497-250MG A109497-5G
76869-644EA 43 USD
76869-644 76869-646 76869-648 76869-650
4,5-Difluorobenzene-1,2-diol 97%
4,5-Difluorobenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₆H₄F₂O₂
MDL Number: MFCD08458183
CAS Number: 147300-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR