N-(3-Bromophenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A109206-250MG A109206-1G A109206-5G
76869-632EA 8.49 USD
76869-632 76869-634 77884-994 77884-995
N-(3-Bromophenyl)methanesulfonamide 98%
N-(3-Bromophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
MDL Number: MFCD01213915
CAS Number: 83922-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR