8-Bromoquinazolin-4-amine 95%

Supplier: Ambeed
Warning

A108819-1G A108819-250MG
76869-612EA 62 USD
76869-612 76869-614
8-Bromoquinazolin-4-amine 95%
8-Bromoquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MDL Number: MFCD09027697
CAS Number: 1260657-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR