Boc-D-Asp-OBzl 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-aspartic acid 1-benzyl ester

A108809-1G A108809-250MG A108809-5G A108809-25G A108809-10G
76869-606EA 15.92 USD
76869-606 76869-608 76869-610 77549-098 77610-860
Boc-D-Asp-OBzl 98%
Boc-D-Asp-OBzl

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆
MDL Number: MFCD00038264
CAS Number: 92828-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR