(E)-3-(6-Aminopyridin-3-yl)acrylic acid 97%
Supplier: Ambeed
|
Warning
|
A108631-1G
A108631-250MG
A108631-5G
A108631-25G
76869-600EA
10
USD
76869-600
76869-602
76869-604
77882-913
(E)-3-(6-Aminopyridin-3-yl)acrylic acid 97%
(E)-3-(6-Aminopyridin-3-yl)acrylic acid
Precaución: for Research Use Only
Formula:
C₈H₈N₂O₂ |
MDL Number:
MFCD14585070 CAS Number: 167837-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...