Dimethyl 2,5-dibromohexanedioate 97%

Supplier: Ambeed
Danger

A108566-100MG A108566-10G A108566-1G A108566-250MG A108566-25G A108566-5G
76869-588EA 5.31 USD
76869-588 76869-590 76869-592 76869-594 76869-596 76869-598
Dimethyl 2,5-dibromohexanedioate 97%
Dimethyl 2,5-dibromohexanedioate

Precaución: for Research Use Only

Formula: C₈H₁₂Br₂O₄
MW: 331.98 g/mol
MDL Number: MFCD00043758
CAS Number: 868-72-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR