4-Bromo-N,N-dimethyl-2-nitroaniline 98%

Supplier: Ambeed
Warning

A108460-1G A108460-25G A108460-5G
76869-582EA 26 USD
76869-582 76869-584 76869-586
4-Bromo-N,N-dimethyl-2-nitroaniline 98%
4-Bromo-N,N-dimethyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O₂
MDL Number: MFCD12546467
CAS Number: 829-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR