Benzo[1,2-b:4,5-b']dithiophene-4,8-dione 97%

Supplier: Ambeed
Warning

A108386-100G A108386-1G A108386-250MG A108386-25G A108386-5G A108386-10G
76869-566EA 727 USD
76869-566 76869-568 76869-570 76869-572 76869-574 77570-488
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione 97%
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione

Precaución: for Research Use Only

Formula: C₁₀H₄O₂S₂
MDL Number: MFCD01927240
CAS Number: 32281-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR