AS-605240 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Quinoxalin-6-ylmethylene)thiazolidine-2,4-dione

A108127-10MG A108127-1MG A108127-50MG A108127-5MG
76869-546EA 46 USD
76869-546 76869-548 76869-550 76869-552
AS-605240 98%
AS-605240

Precaución: for Research Use Only

Formula: C₁₂H₇N₃O₂S
MDL Number: MFCD11114066
CAS Number: 648450-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR