4-Iodo-1-methylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-1-methyl-1H-imidazole

A108011-100MG A108011-10G A108011-1G A108011-250MG A108011-25G A108011-5G
76869-534EA 5.31 USD
76869-534 76869-536 76869-538 76869-540 76869-542 76869-544
4-Iodo-1-methylimidazole 98%
4-Iodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD01632214
CAS Number: 71759-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR