4-Chloroindole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-1H-indole-3-carbaldehyde

A107815-250MG A107815-5G
76869-522EA 10.61 USD
76869-522 76869-524
4-Chloroindole-3-carbaldehyde 98%
4-Chloroindole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD05864704
CAS Number: 876-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR