2,4,6-Trihydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A107723-10G A107723-1G A107723-25G
76869-514EA 28.65 USD
76869-514 76869-516 76869-518
2,4,6-Trihydroxybenzoic acid 95%
2,4,6-Trihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₅
MW: 170.12 g/mol
MDL Number: MFCD00002453
CAS Number: 83-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR