4-Bromo-1H-pyrrole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A107346-100MG A107346-1G A107346-250MG A107346-5G
76869-500EA 18 USD
76869-500 76869-502 76869-504 76869-506
4-Bromo-1H-pyrrole-2-carbonitrile 95%
4-Bromo-1H-pyrrole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₃BrN₂
MDL Number: MFCD20502410
CAS Number: 1221435-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR