4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Ethyl-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl
A107269-100G
A107269-10G
A107269-1G
A107269-25G
A107269-5G
76869-490EA
63
USD
76869-490
76869-492
76869-494
76869-496
76869-498
4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl 98%
4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl
Precaución: for Research Use Only
Formula:
C₂₃H₃₀ MW: 306.49 g/mol |
MDL Number:
MFCD09751090 CAS Number: 84540-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...