2-(3-Fluoro-2-nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A107189-1G A107189-5G
76869-488EA 65 USD
76869-488 77882-904
2-(3-Fluoro-2-nitrophenyl)acetic acid 97%
2-(3-Fluoro-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD13192267
CAS Number: 872141-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR