1,4-Dimethyl-1H-pyrazol-3-amine 98%

Supplier: Ambeed
Warning

A106669-100MG A106669-250MG
76869-462EA 87 USD
76869-462 76869-464
1,4-Dimethyl-1H-pyrazol-3-amine 98%
1,4-Dimethyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₅H₉N₃
MW: 111.14 g/mol
MDL Number: MFCD11215414
CAS Number: 85485-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR