2-Aminobenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole

A106367-100G A106367-10G A106367-25G A106367-500G A106367-5G A106367-1000G
76869-448EA 37.14 USD
76869-448 76869-450 76869-452 76869-454 76869-456 77610-838
2-Aminobenzimidazole 98%
2-Aminobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00005596
CAS Number: 934-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR