(1R,2R)-2-Fluorocyclopropanecarboxylic acid 98%

Supplier: Ambeed
Danger

A105139-1G A105139-5G A105139-250MG
76869-382EA 43.5 USD
76869-382 77570-312 77570-310
(1R,2R)-2-Fluorocyclopropanecarboxylic acid 98%
(1R,2R)-2-Fluorocyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅FO₂
MW: 104.08 g/mol
MDL Number: MFCD19238080
CAS Number: 127199-13-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR