tert-Butyl (4-(chlorosulfonyl)phenyl)carbamate 98%

Supplier: Ambeed
Danger

A104979-100MG A104979-1G A104979-5G A104979-250MG
76869-374EA 12 USD
76869-374 76869-376 76869-378 77570-292
tert-Butyl (4-(chlorosulfonyl)phenyl)carbamate 98%
tert-Butyl (4-(chlorosulfonyl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₄S
MW: 291.751 g/mol
MDL Number: MFCD10697736
CAS Number: 269747-25-3
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR