Methyl 3-Amino-5-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-bromo-2-methylbenzoic acid methyl ester, Methyl 3-amino-5-bromo-2-methylbenzoate

A104752-1G A104752-5G A104752-100G A104752-10G A104752-25G
76869-348EA 6.37 USD
76869-348 76869-350 77548-996 77570-278 77570-280
Methyl 3-Amino-5-bromo-2-methylbenzoate 98%
Methyl 3-Amino-5-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD08690071
CAS Number: 1000342-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR