5-Bromo-6-methoxybenzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methoxy-1H-benzo[d]imidazole

A104386-1G A104386-250MG
76869-330EA 166.57 USD
76869-330 76869-332
5-Bromo-6-methoxybenzimidazole 97%
5-Bromo-6-methoxybenzimidazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
MDL Number: MFCD18783164
CAS Number: 1008361-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR