(4-Bromothiophen-2-yl)methanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A104205-100MG
A104205-250MG
76869-312EA
70
USD
76869-312
76869-314
(4-Bromothiophen-2-yl)methanamine hydrochloride 97%
(4-Bromothiophen-2-yl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₇BrClNS |
MDL Number:
MFCD09878584 CAS Number: 1171697-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...