(5,6,7,8-Tetrahydronaphthalen-1-yl)methanol 97%

Supplier: Ambeed
Warning

A104128-100MG A104128-250MG
76869-300EA 46.68 USD
76869-300 76869-302
(5,6,7,8-Tetrahydronaphthalen-1-yl)methanol 97%
(5,6,7,8-Tetrahydronaphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD16037293
CAS Number: 41790-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR