Imipenem monohydrate 98%

Supplier: Ambeed
Danger

Synonyms: (5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate

A103901-250MG A103901-100MG A103901-1G
76869-292EA 7.43 USD
76869-292 77670-506 77882-890 77882-891
Imipenem monohydrate 98%
Imipenem monohydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₄S
MW: 317.36 g/mol
MDL Number: MFCD09753321
CAS Number: 74431-23-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR