5-Bromo-4-methylthiophene-2-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-4-methylthiophen-2-yl)boronic acid

A103896-100MG A103896-1G A103896-250MG A103896-5G
76869-284EA 37.14 USD
76869-284 76869-286 76869-288 76869-290
5-Bromo-4-methylthiophene-2-boronic acid 95%
5-Bromo-4-methylthiophene-2-boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BBrO₂S
MW: 220.88 g/mol
MDL Number: MFCD12913941
CAS Number: 1217501-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR