CX-4945 98%

Supplier: Ambeed
Warning

Synonyms: Silmitasertib, 5-((3-Chlorophenyl)amino)benzo[c][2,6]naphthyridine-8-carboxylic acid

A103748-1MG A103748-250MG A103748-50MG A103748-5MG A103748-100MG
76869-270EA 6.37 USD
76869-270 76869-272 76869-274 76869-276 77570-216
CX-4945 98%
CX-4945

Precaución: for Research Use Only

Formula: C₁₉H₁₂ClN₃O₂
MW: 349.8 g/mol
MDL Number: MFCD13184796
CAS Number: 1009820-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR