(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid 98%

Supplier: Ambeed
Warning

A103726-1000G A103726-500G A103726-100G A103726-25G A103726-5G A103726-10G
76869-266EA 413 USD
76869-266 76869-268 77610-792 77610-794 77670-502 77882-886
(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid 98%
(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₆
MDL Number: MFCD00063182
CAS Number: 78663-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR