Methyl 3-cyano-1H-indole-7-carboxylate 95%

Supplier: Ambeed
Warning

A103437-1G A103437-250MG
76869-258EA 42 USD
76869-258 76869-260
Methyl 3-cyano-1H-indole-7-carboxylate 95%
Methyl 3-cyano-1H-indole-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₂
MDL Number: MFCD11976634
CAS Number: 443144-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR