Ranolazine 98%

Supplier: Ambeed
Warning

A103291-100MG A103291-250MG A103291-5G A103291-100G A103291-10G
76869-248EA 5 USD
76869-248 76869-250 76869-252 77884-935 77884-936
Ranolazine 98%
Ranolazine

Precaución: for Research Use Only

Formula: C₂₄H₃₃N₃O₄
MDL Number: MFCD00864690
CAS Number: 95635-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR