Bicyclo[1.1.1]pentane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A103228-50MG A103228-1G A103228-250MG A103228-100MG A103228-5G
76869-246EA 61.54 USD
76869-246 77548-950 77548-952 77570-184 77570-186
Bicyclo[1.1.1]pentane-1-carboxylic acid 98%
Bicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.127 g/mol
MDL Number: MFCD19228106
CAS Number: 22287-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR