3-Fluoro-4-nitrobenzylalcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-nitrophenyl)methanol

A103186-1G A103186-250MG A103186-5G A103186-25G
76869-240EA 18 USD
76869-240 76869-242 76869-244 77670-490
3-Fluoro-4-nitrobenzylalcohol 98%
3-Fluoro-4-nitrobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD03094236
CAS Number: 503315-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR