6-(Dimethylamino)purine 97%

Supplier: Ambeed
Warning

A103153-100MG A103153-10G A103153-1G A103153-1MG A103153-5G A103153-25G
76869-230EA 17 USD
76869-230 76869-232 76869-234 76869-236 76869-238 77884-934
6-(Dimethylamino)purine 97%
6-(Dimethylamino)purine

Precaución: for Research Use Only

Formula: C₇H₉N₅
MDL Number: MFCD00005573
CAS Number: 938-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR