2,4-Dichloro-6-iodoaniline 98%

Supplier: Ambeed
Warning

A102977-1G A102977-5G A102977-25G A102977-10G
76869-212EA 6 USD
76869-212 76869-214 77570-174 77670-480
2,4-Dichloro-6-iodoaniline 98%
2,4-Dichloro-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂IN
MDL Number: MFCD08533279
CAS Number: 697-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR