5-Bromo-3-nitrobenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A102413-100MG A102413-1G A102413-250MG
76869-190EA 41 USD
76869-190 76869-192 76869-194
5-Bromo-3-nitrobenzene-1,2-diamine 95%
5-Bromo-3-nitrobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MDL Number: MFCD09817647
CAS Number: 84752-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR