4,6-Dichloro-5-iodo-2-methylpyrimidine 97%

Supplier: Ambeed
Warning

A868560-1G A868560-100MG A868560-250MG A868560-50MG
76845-534EA 683.22 USD
76845-534 77244-568 77244-570 77244-572
4,6-Dichloro-5-iodo-2-methylpyrimidine 97%
4,6-Dichloro-5-iodo-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂IN₂
MW: 288.90 g/mol
MDL Number: MFCD21743354
CAS Number: 111079-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR