Diisopropyl azodicarboxylate 98%

Supplier: Ambeed
Danger

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

A848539-25G A848539-100G A848539-500G
76845-472EA 23 USD
76845-472 77552-588 77552-590
Diisopropyl azodicarboxylate 98%
Diisopropyl azodicarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
MDL Number: MFCD00008875
CAS Number: 2446-83-5
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR