Avanafil 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(3-Chloro-4-methoxybenzyl)amino]-2-[(2S)-2-(hydroxymethyl)pyrrolidin-1-yl]-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide, Avanfil, (S)-4-((3-Chloro-4-methoxybenzyl)amino)-2-(2-(hydroxymethyl)pyrrolidin-1-yl)-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide

A803364-1G A803364-100MG A803364-10MG A803364-250MG A803364-50MG A803364-25G A803364-5G
76845-316EA 42.44 USD
76845-316 77171-380 77171-382 77171-384 77171-386 77684-749 77684-750
Avanafil 98%
Avanafil

Precaución: For research use only.

Formula: C₂₃H₂₆ClN₇O₃
MW: 483.951 g/mol
MDL Number: MFCD11977961
CAS Number: 330784-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR