6-Bromo-3-ethyl-2-methoxyquinoline 95%

Supplier: Ambeed
Warning

A763863-1G A763863-100MG A763863-250MG
76845-134EA 212.18 USD
76845-134 77036-194 77036-196
6-Bromo-3-ethyl-2-methoxyquinoline 95%
6-Bromo-3-ethyl-2-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrNO
MW: 266.13 g/mol
MDL Number: MFCD09029947
CAS Number: 409346-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR