2,5-Diaminobenzenesulfonic acid 95%

Supplier: Ambeed
Warning

A757615-25G A757615-100G A757615-10G A757615-500G
76845-110EA 14 USD
76845-110 77118-202 77118-204 77118-206 77118-208
2,5-Diaminobenzenesulfonic acid 95%
2,5-Diaminobenzenesulphonic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃S
MDL Number: MFCD00007904
CAS Number: 88-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR