4-((6-((3-Chloropropanoyl)oxy)hexyl)oxy)benzoic acid 97%

Supplier: Ambeed
Warning

A731995-1G A731995-100MG A731995-250MG
76845-014EA 128.37 USD
76845-014 77018-910 77018-912
4-((6-((3-Chloropropanoyl)oxy)hexyl)oxy)benzoic acid 97%
4-((6-((3-Chloropropanoyl)oxy)hexyl)oxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₁ClO₅
MW: 328.79 g/mol
MDL Number: MFCD27920914
CAS Number: 182922-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR