Valdecoxib 99+%

Supplier: Ambeed
Warning

Synonyms: 4-(5-methyl-3-phenylisoxazol-4-yl)benzenesulphonamide

A727456-1G A727456-100G A727456-10MG A727456-250MG A727456-25G A727456-50MG A727456-5G
76844-994EA 16.98 USD
76844-994 77035-950 77035-952 77035-954 77035-956 77035-958 77035-960
Valdecoxib 99+%
Valdecoxib

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂O₃S
MW: 314.36 g/mol
MDL Number: MFCD00950568
CAS Number: 181695-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR