2-(Methylthio)pyrimidine-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A725540-1G A725540-250MG A725540-25G A725540-5G
76844-986EA 41 USD
76844-986 77035-918 77035-920 77035-922
2-(Methylthio)pyrimidine-5-carboxylic acid 98%
2-(Methylthio)pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂S
MDL Number: MFCD07394096
CAS Number: 110099-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR