Tetrahydropyran-3-methanol 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)tetrahydro-2H-pyran, (Tetrahydropyran-3-yl)methanol

A711065-1G A711065-100MG A711065-250MG A711065-5G
76844-916EA 111.4 USD
76844-916 77198-904 77198-906 77198-908
Tetrahydropyran-3-methanol 95%
Tetrahydropyran-3-methanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.1583 g/mol
MDL Number: MFCD08235011
CAS Number: 14774-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR