3-Bromo-6-methoxyquinoline 96%

Supplier: Ambeed
Warning

A709682-1G A709682-100MG A709682-5G A709682-250MG
76844-908EA 90 USD
76844-908 76981-160 76981-162 77673-948
3-Bromo-6-methoxyquinoline 96%
3-Bromo-6-methoxyquinoline

Precaución: For research use only

Formula: C₁₀H₈BrNO
MDL Number: MFCD09260428
CAS Number: 14036-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR