2-Chloro-5-nitroaniline 95%

Supplier: Ambeed
Danger

A708113-25G A708113-100G
76844-902EA 14 USD
76844-902 77673-944
2-Chloro-5-nitroaniline 95%
2-Chloro-5-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 119
MDL Number: MFCD00007668
CAS Number: 6283-25-6
UN: 2237
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR