(4-Aminophenoxy)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Aminophenoxy)acetic acid

A698244-1G A698244-100MG A698244-250MG A698244-5G
76844-842EA 118.83 USD
76844-842 77243-434 77243-436 77243-438
(4-Aminophenoxy)acetic acid 95%
(4-Aminophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD00224890
CAS Number: 2298-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR