2-Chloropyrazine 98%

Supplier: Ambeed
Danger

Synonyms: Chloropyrazine

A697814-25G A697814-100G
76844-838EA 5.31 USD
76844-838 77614-150
2-Chloropyrazine 98%
2-Chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
MDL Number: MFCD00006124
CAS Number: 14508-49-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR